2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde 98%
Lieferant: Apollo Scientific
Synonyme:
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde
PC302457-5G
PC302457-1G
APOSPC302457-5GEA
116
EUR
APOSPC302457-5G
APOSPC302457-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde
Formel:
C₉H₆F₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201033 CAS-Nummer: 1373920-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...