2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde

PC302457-5G PC302457-1G
APOSPC302457-5GEA 116 EUR
APOSPC302457-5G APOSPC302457-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde
Formel: C₉H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22201033
CAS-Nummer: 1373920-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR