2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide 98%

Lieferant: Apollo Scientific
Gefahr

PC302460-1G
APOSPC302460-1GEA 0 EUR
APOSPC302460-1G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide
Formel: C₉H₇BrF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD19687198
CAS-Nummer: 1323966-23-9
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR