(2-Methoxy-3-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

PC302462-1G
APOSPC302462-1GEA 116 EUR
APOSPC302462-1G
(2-Methoxy-3-(trifluormethyl)phenyl)essigsäure 98%
(2-Methoxy-3-(trifluormethyl)phenyl)essigsäure
Formel: C₁₀H₉F₃O₃
Molecular Weight: 234,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832317
CAS-Nummer: 1017778-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR