2-Methoxy-4-(trifluoromethoxy)benzamide

Lieferant: Apollo Scientific

PC302463-10G PC302463-5G
APOSPC302463-10GEA 395 EUR
APOSPC302463-10G APOSPC302463-5G
2-Methoxy-4-(trifluoromethoxy)benzamide
2-Methoxy-4-(trifluoromethoxy)benzamide
Formel: C₉H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660245
CAS-Nummer: 886500-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR