2-Methoxy-4-(trifluoromethoxy)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC302464-10G PC302464-5G
APOSPC302464-10GEA 0 EUR
APOSPC302464-10G APOSPC302464-5G
2-Methoxy-4-(trifluoromethoxy)benzyl alcohol
2-Methoxy-4-(trifluoromethoxy)benzyl alcohol
Formel: C₉H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04115969
CAS-Nummer: 886500-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR