2-Methoxy-5-(pentafluorothio)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxy-5-(pentafluorosulphanyl)benzoic acid

PC302467-1G PC302467-5G PC302467-10G
APOSPC302467-1GEA 348 EUR
APOSPC302467-1G APOSPC302467-5G APOSPC302467-10G
2-Methoxy-5-(pentafluorothio)benzoic acid 97%
2-Methoxy-5-(pentafluorothio)benzoic acid
Formel: C₈H₇F₅O₃S
Molecular Weight: 278.196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652486
CAS-Nummer: 1448319-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR