2-Methyl-3-(trifluormethyl)anisol 98%
Lieferant: Apollo Scientific
PC302469-1G
APOSPC302469-1GEA
116
EUR
APOSPC302469-1G
2-Methyl-3-(trifluormethyl)anisol 98%
2-Methyl-3-(trifluormethyl)anisol
Formel:
C₉H₉F₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832284 CAS-Nummer: 1017778-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...