2-Methyl-5-(trifluormethyl)phenylacetonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302475-1G
PC302475-5G
PC302475-10G
APOSPC302475-1GEA
0
EUR
APOSPC302475-1G
APOSPC302475-5G
APOSPC302475-10G
2-Methyl-5-(trifluormethyl)phenylacetonitril 98%
2-Methyl-5-(trifluormethyl)phenylacetonitril
Formel:
C₁₀H₈F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832285 CAS-Nummer: 1000513-69-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...