2-Methyl-5-(trifluormethyl)phenylacetonitril 98%

Lieferant: Apollo Scientific
Gefahr

PC302475-1G PC302475-5G PC302475-10G
APOSPC302475-1GEA 0 EUR
APOSPC302475-1G APOSPC302475-5G APOSPC302475-10G
2-Methyl-5-(trifluormethyl)phenylacetonitril 98%
2-Methyl-5-(trifluormethyl)phenylacetonitril
Formel: C₁₀H₈F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD09832285
CAS-Nummer: 1000513-69-8
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR