3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid

PC302486-1G
APOSPC302486-1GEA 224 EUR
APOSPC302486-1G
3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid 98%
3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid
Formel: C₁₀H₁₀ClFO₃
Molecular Weight: 232,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687213
CAS-Nummer: 1323966-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR