3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%

Lieferant: Apollo Scientific
Achtung

PC302487-1G
APOSPC302487-1GEA 116 EUR
APOSPC302487-1G
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid
Formel: C₁₁H₁₂ClFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19687220
CAS-Nummer: 1323966-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR