3-(4-Fluoro-3,5-dimethylphenyl)propionic acid 98%

Lieferant: Apollo Scientific

PC302489-1G
APOSPC302489-1GEA 342 EUR
APOSPC302489-1G
3-(4-Fluoro-3,5-dimethylphenyl)propionic acid 98%
3-(4-Fluoro-3,5-dimethylphenyl)propionic acid
Formel: C₁₁H₁₃FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832377
CAS-Nummer: 1017779-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR