3-(Trifluoromethoxy)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302493-1G PC302493-5G PC302493-250MG PC302493-25G
APOSPC302493-1GEA 0 EUR
APOSPC302493-1G APOSPC302493-5G APOSPC302493-250MG APOSPC302493-25G
3-(Trifluoromethoxy)-DL-phenylalanine
3-(Trifluoromethoxy)-DL-phenylalanine
Formel: C₁₀H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04116035
CAS-Nummer: 439587-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR