3,6-Dichlor-2-fluorbenzamid 97%

Lieferant: Apollo Scientific

PC302495-5G PC302495-10G PC302495-1G
APOSPC302495-5GEA 136 EUR
APOSPC302495-5G APOSPC302495-10G APOSPC302495-1G
3,6-Dichlor-2-fluorbenzamid 97%
3,6-Dichlor-2-fluorbenzamid
Formel: C₇H₄Cl₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09025384
CAS-Nummer: 916420-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR