3,6-Dichlor-2-fluorbenzylalkohol 98%

Lieferant: Apollo Scientific

PC302496-5G
APOSPC302496-5GEA 212 EUR
APOSPC302496-5G
3,6-Dichlor-2-fluorbenzylalkohol 98%
3,6-Dichlor-2-fluorbenzylalkohol
Formel: C₇H₅Cl₂FO
Molecular Weight: 195,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025388
CAS-Nummer: 916420-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR