5,6-Difluoroxindol 95+%

Lieferant: Apollo Scientific

Synonyme: 5,6-Difluoroindolin-2-one

PC3025-1G
APOSPC3025-1GEA 97.3 EUR
APOSPC3025-1G
5,6-Difluoroxindol 95+%
5,6-Difluoroxindol
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075216
CAS-Nummer: 71294-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR