3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid, Benzenepropanoic acid, 2-fluoro-5-methyl-4-(trifluoromethyl)-

PC302500-1G
APOSPC302500-1GEA 166 EUR
APOSPC302500-1G
3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid 98%
3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid
Formel: C₁₁H₁₀F₄O₂
Molecular Weight: 250,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687202
CAS-Nummer: 1323966-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR