3-(3-(Difluormethoxy)phenyl)propionsäure 98%

Lieferant: Apollo Scientific

PC302504-1G
APOSPC302504-1GEA 114 EUR
APOSPC302504-1G
3-(3-(Difluormethoxy)phenyl)propionsäure 98%
3-(3-(Difluormethoxy)phenyl)propionsäure
Formel: C₁₀H₁₀F₂O₃
Molecular Weight: 216,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519314
CAS-Nummer: 1092460-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR