3-(3-Chlor-5-(trifluormethoxy)phenyl)propionsäure 98%

Lieferant: Apollo Scientific

PC302506-1G
APOSPC302506-1GEA 171 EUR
APOSPC302506-1G
3-(3-Chlor-5-(trifluormethoxy)phenyl)propionsäure 98%
3-(3-Chlor-5-(trifluormethoxy)phenyl)propionsäure
Formel: C₁₀H₈ClF₃O₃
Molecular Weight: 268.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025398
CAS-Nummer: 916420-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR