3-(3-Methyl-5-(trifluormethoxy)phenyl)propionsäure 98%

Lieferant: Apollo Scientific

PC302510-1G
APOSPC302510-1GEA 171 EUR
APOSPC302510-1G
3-(3-Methyl-5-(trifluormethoxy)phenyl)propionsäure 98%
3-(3-Methyl-5-(trifluormethoxy)phenyl)propionsäure
Formel: C₁₁H₁₁F₃O₃
Molecular Weight: 248.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025376
CAS-Nummer: 916420-57-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR