3-(4-(Difluormethoxy)phenyl)propionsäure 97%

Lieferant: Apollo Scientific

PC302513-1G
APOSPC302513-1GEA 116 EUR
APOSPC302513-1G
3-(4-(Difluormethoxy)phenyl)propionsäure 97%
3-(4-(Difluormethoxy)phenyl)propionsäure
Formel: C₁₀H₁₀F₂O₃
Schmelzpunkt: 66…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09863355
CAS-Nummer: 1000932-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR