3-(4-Fluor-3-(trifluormethoxy)phenyl)propionsäure 98%

Lieferant: Apollo Scientific

PC302515-1G
APOSPC302515-1GEA 109 EUR
APOSPC302515-1G
3-(4-Fluor-3-(trifluormethoxy)phenyl)propionsäure 98%
3-(4-Fluor-3-(trifluormethoxy)phenyl)propionsäure
Formel: C₁₀H₈F₄O₃
Molecular Weight: 252,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09999449
CAS-Nummer: 1240256-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR