3-Chloro-2-fluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302521-1G
PC302521-5G
PC302521-10G
APOSPC302521-1GEA
116
EUR
APOSPC302521-1G
APOSPC302521-5G
APOSPC302521-10G
3-Chloro-2-fluoro-DL-phenylalanine
3-Chloro-2-fluoro-DL-phenylalanine
Formel:
C₉H₉ClFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116015 CAS-Nummer: 1259993-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...