3-Chloro-2-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302521-1G PC302521-5G PC302521-10G
APOSPC302521-1GEA 116 EUR
APOSPC302521-1G APOSPC302521-5G APOSPC302521-10G
3-Chloro-2-fluoro-DL-phenylalanine
3-Chloro-2-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116015
CAS-Nummer: 1259993-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR