3-Chloro-5-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

PC302532-5G PC302532-1G
APOSPC302532-5GEA 0 EUR
APOSPC302532-5G APOSPC302532-1G
3-Chloro-5-(trifluoromethyl)benzamide 97%
3-Chloro-5-(trifluoromethyl)benzamide
Formel: C₈H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD04115853
CAS-Nummer: 886497-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR