3-Chlor-5-fluor-4-methoxybenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC302537-5G
APOSPC302537-5GEA 154 EUR
APOSPC302537-5G
3-Chlor-5-fluor-4-methoxybenzoesäure 98%
3-Chloro-5-fluoro-4-methoxybenzoic acid
Formel: C₈H₆ClFO₃
Molecular Weight: 204,59 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115864
CAS-Nummer: 886497-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR