3-Chloro-5-fluoro-4-methoxyphenylacetonitrile 98%
Lieferant: Apollo Scientific
PC302540-1G
APOSPC302540-1GEA
159
EUR
APOSPC302540-1G
3-Chloro-5-fluoro-4-methoxyphenylacetonitrile 98%
3-Chloro-5-fluoro-4-methoxyphenylacetonitrile
Formel:
C₉H₇ClFNO Molecular Weight: 199,609 g/mol Schmelzpunkt: 37…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832263 CAS-Nummer: 1000540-93-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...