3-Chlor-5-fluorbenzamid

Lieferant: Apollo Scientific

PC302541-5G
APOSPC302541-5GEA 59.6 EUR
APOSPC302541-5G
3-Chlor-5-fluorbenzamid
3-Chlor-5-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57
Lagertemperatur: Raumtemperatur
MDL: MFCD03407964
CAS-Nummer: 874781-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR