3-Chloro-5-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(3-chloro-5-fluorophenyl)propanoic acid

PC302542-1G
APOSPC302542-1GEA 116 EUR
APOSPC302542-1G
3-Chloro-5-fluoro-DL-phenylalanine
3-Chloro-5-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Molecular Weight: 217,625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115837
CAS-Nummer: 1259985-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR