3-Fluor-4-(trifluormethoxy)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC302551-1G PC302551-5G PC302551-10G
APOSPC302551-1GEA 0 EUR
APOSPC302551-1G APOSPC302551-5G APOSPC302551-10G
3-Fluor-4-(trifluormethoxy)benzylbromid
3-Fluor-4-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660205
CAS-Nummer: 886499-04-3
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR