3-Fluor-5-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302554-1G PC302554-5G PC302554-10G
APOSPC302554-1GEA 0 EUR
APOSPC302554-1G APOSPC302554-5G APOSPC302554-10G
3-Fluor-5-(trifluormethoxy)benzylbromid 98%
3-Fluor-5-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD18783226
CAS-Nummer: 86256-51-1
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR