3-Fluor-5-(trifluormethoxy)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302554-1G
PC302554-5G
PC302554-10G
APOSPC302554-1GEA
0
EUR
APOSPC302554-1G
APOSPC302554-5G
APOSPC302554-10G
3-Fluor-5-(trifluormethoxy)benzylbromid 98%
3-Fluor-5-(trifluormethoxy)benzylbromid
Formel:
C₈H₅BrF₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18783226 CAS-Nummer: 86256-51-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...