3-Fluoro-5-methyl-DL-phenylglycine

Lieferant: Apollo Scientific

PC302556-5G
APOSPC302556-5GEA 0 EUR
APOSPC302556-5G
3-Fluoro-5-methyl-DL-phenylglycine
3-Fluoro-5-methyl-DL-phenylglycine
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662466
CAS-Nummer: 199327-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR