3-Methoxy-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302558-1G PC302558-5G PC302558-10G
APOSPC302558-1GEA 159 EUR
APOSPC302558-1G APOSPC302558-5G APOSPC302558-10G
3-Methoxy-2-(trifluormethyl)benzylbromid 98%
3-Methoxy-2-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18398595
CAS-Nummer: 1261828-76-5
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR