4-Chlor-2-(trifluormethoxy)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC302566-1G PC302566-5G PC302566-10G
APOSPC302566-1GEA 0 EUR
APOSPC302566-1G APOSPC302566-5G APOSPC302566-10G
4-Chlor-2-(trifluormethoxy)benzylbromid 97%
4-Chlor-2-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18393833
CAS-Nummer: 1261606-06-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR