4'-Chlor-2'-fluor-3'-methoxyacetophenon 98%

Lieferant: Apollo Scientific

PC302568-1G PC302568-5G PC302568-10G
APOSPC302568-1GEA 116 EUR
APOSPC302568-1G APOSPC302568-5G APOSPC302568-10G
4'-Chlor-2'-fluor-3'-methoxyacetophenon 98%
4'-Chlor-2'-fluor-3'-methoxyacetophenon
Formel: C₉H₈ClFO₂
Molecular Weight: 202.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687210
CAS-Nummer: 1323966-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR