4-Chloro-2-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC302572-1G PC302572-5G PC302572-10G
APOSPC302572-1GEA 357 EUR
APOSPC302572-1G APOSPC302572-5G APOSPC302572-10G
4-Chloro-2-fluoro-DL-phenylalanine
4-Chloro-2-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115957
CAS-Nummer: 754944-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR