4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC302573-25G PC302573-5G PC302573-50G
APOSPC302573-25GEA 449 EUR
APOSPC302573-25G APOSPC302573-5G APOSPC302573-50G
4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%
4-Chloro-3-(trifluoromethylthio)benzyl bromide
Formel: C₈H₅BrClF₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD06660314
CAS-Nummer: 886503-38-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR