4-Chloro-3,5-difluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302576-1G
APOSPC302576-1GEA 0 EUR
APOSPC302576-1G
4-Chloro-3,5-difluoro-DL-phenylalanine
4-Chloro-3,5-difluoro-DL-phenylalanine
Formel: C₉H₈ClF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23795327
CAS-Nummer: 1706431-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR