4-Chloro-3,5-difluorophenylacetic acid 98%

Lieferant: Apollo Scientific

PC302577-1G PC302577-5G PC302577-10G
APOSPC302577-1GEA 224 EUR
APOSPC302577-1G APOSPC302577-5G APOSPC302577-10G
4-Chloro-3,5-difluorophenylacetic acid 98%
4-Chloro-3,5-difluorophenylacetic acid
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206.574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09925051
CAS-Nummer: 1000566-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR