4-Chloro-3-ethoxy-2-fluorobenzamide 97%

Lieferant: Apollo Scientific

Synonyme: 4-chloro-3-ethoxy-2-fluorobenzamide

PC302578-1G
APOSPC302578-1GEA 116 EUR
APOSPC302578-1G
4-Chloro-3-ethoxy-2-fluorobenzamide 97%
4-Chloro-3-ethoxy-2-fluorobenzamide
Formel: C₉H₉ClFNO₂
Molecular Weight: 217,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687217
CAS-Nummer: 1323966-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR