(4-Ethoxy-2,3-difluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific

PC302583-1G
APOSPC302583-1GEA 136 EUR
APOSPC302583-1G
(4-Ethoxy-2,3-difluorphenyl)essigsäure 97%
(4-Ethoxy-2,3-difluorphenyl)essigsäure
Formel: C₁₀H₁₀F₂O₃
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258707
CAS-Nummer: 1017779-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR