4-Ethoxy-2-fluoroaniline 98%
Lieferant: Apollo Scientific
PC302585-1G
APOSPC302585-1GEA
119
EUR
APOSPC302585-1G
4-Ethoxy-2-fluoroaniline 98%
4-Ethoxy-2-fluoroaniline
Formel:
C₈H₁₀FNO Molecular Weight: 155,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053973 CAS-Nummer: 470702-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...