4-Fluor-2-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-4-fluoro-2-(trifluoromethoxy)benzene

PC302591-1G
APOSPC302591-1GEA 161 EUR
APOSPC302591-1G
4-Fluor-2-(trifluormethoxy)benzylbromid 98%
4-Fluor-2-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Molecular Weight: 273,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687175
CAS-Nummer: 1323966-22-8
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR