4-Fluoro-2-methoxycinnamic acid 97%
Lieferant: Apollo Scientific
PC302592-1G
APOSPC302592-1GEA
116
EUR
APOSPC302592-1G
4-Fluoro-2-methoxycinnamic acid 97%
4-Fluoro-2-methoxycinnamic acid
Formel:
C₁₀H₉FO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03002810 CAS-Nummer: 682804-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...