4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde

PC302596-10G PC302596-5G PC302596-1G
APOSPC302596-10GEA 371 EUR
APOSPC302596-10G APOSPC302596-5G APOSPC302596-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde
Formel: C₉H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD22201063
CAS-Nummer: 1373921-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR