4-Methoxy-2-(trifluormethyl)benzolsulfonylchlorid 98%
Lieferant: Apollo Scientific
PC302599-1G
PC302599-5G
PC302599-10G
APOSPC302599-1GEA
116
EUR
APOSPC302599-1G
APOSPC302599-5G
APOSPC302599-10G
4-Methoxy-2-(trifluormethyl)benzolsulfonylchlorid 98%
4-Methoxy-2-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₈H₆ClF₃O₃S Schmelzpunkt: 57…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519323 CAS-Nummer: 775288-85-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...