4-Methoxy-3-(trifluoromethoxy)benzamide 98%

Lieferant: Apollo Scientific

PC302602-1G PC302602-5G PC302602-10G
APOSPC302602-1GEA 0 EUR
APOSPC302602-1G APOSPC302602-5G APOSPC302602-10G
4-Methoxy-3-(trifluoromethoxy)benzamide 98%
4-Methoxy-3-(trifluoromethoxy)benzamide
Formel: C₉H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18398425
CAS-Nummer: 1261646-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR