4-Methoxy-3-(trifluormethoxy)benzylbromid

Lieferant: Apollo Scientific

PC302603-1G PC302603-5G
APOSPC302603-1GEA 224 EUR
APOSPC302603-1G APOSPC302603-5G
4-Methoxy-3-(trifluormethoxy)benzylbromid
4-Methoxy-3-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18398486
CAS-Nummer: 853771-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR